Learn More
Corning™ Nonsterile Polyester Microplate Sealing Tape
GSA_VA
Catalog No. 07201883 Shop All Corning Products
Change view
Click to view available options
Material:
Polyester
Catalog No. Material
07-201-883 Polyester
1 options
This item is not returnable. View return policy
Catalog No. 07-201-883 Supplier Corning™ Supplier No. 4612
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Provides well-to-well sealing across entire microplate to eliminate edge effect in sensitive ELISA assays. Corning™ Nonsterile Polyester Microplate Sealing Tape is suitable for sealing tissue culture mmicroplates, for short term storage incubation, for transport and the containment of solutions.
  • Easy application and removal for short- and long-term storage
  • Provides tight seal to minimize evaporation and condensation
  • Aluminum sealing tape (Cat. Nos. 6569 and 6570) is suitable for use between -80°C and 150°C, is not transparent, and is pierceable.
  • Breathable sealing tape (Cat. No. 3345) allows gas exchange across the surface.
  • Universal Optical sealing tape (Cat. No. 6575) is suitable for use between -70°C and 100°C, and is transparent.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Material Polyester
Sterility Non-sterile
No. per Case 500/Cs.
No. per Pack 100/Pk.
Product Type Microplate Sealing Tape
Temperature Range (Metric) -40°C to +120°C
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov