Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Polyethylene Caps for Cluster Tube System
GSA_VA

Catalog No. 07200323 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-200-323 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-200-323 Supplier Corning™ Supplier No. 4418
  • PRODUCT PROMOTION
    $83.80
    $36.00 / Case of 120

    Save $47.80

    57% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to fit 1.2mL cluster tubes and cluster tube strips. Corning™ Polyethylene Caps for Cluster Tube System are used with 98-well cluster tube systems.

  • Composed of 96 polypropylene tubes in a standard microplate format.
  • Features 1.2 mL tubes that are available individually or in strips of eight tubes.
  • Polyethylene tube caps are available in 8-cap strips.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type 8-cap Strip
Material Polyethylene
Color Clear
No. per Pack 12/Pk.
No. per Case 120/Cs.
Sterility Sterile
For Use With (Equipment) 1.2 mL 98-well Cluster Tube System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.