Learn More
Corning™ PYREX™ VISTA™ Erlenmeyer Flasks
SCI_ED
Catalog No. S76106A Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
125 mL
250 mL
500 mL
1000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S76106A 25 mL
S76106D 250 mL
S76106B 50 mL
S76106E 500 mL
S76106F 1000 mL
S76106C 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. S76106A Supplier Corning™ Supplier No. 7098025/EMD
This item is not returnable. View return policy
Uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance.
  • Narrow mouth
  • Approximate graduations in durable blue enamel
  • Extra-large marking space
Order Info
Sold in packs of 12.
Compliance
Designed to ASTM™ E-1404, Type 1.
TRUSTED_SUSTAINABILITY
Specifications
Description Blue Enamel graduations; Top style: Beaded rim
Capacity (Metric) 25 mL
Material Borosilicate Glass
Writing Surface Extra Large Blue Marking
Disposable Reusable
Mouth Style Narrow
Sterility Non-sterile