Learn More
Agilent BioTek Corning TC-Treated Plates
GSA_VA
Catalog No. 11120687 Shop All Agilent Technologies Products
Change view
Click to view available options
No. of Positions:
24
96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Positions
11-120-687 96
11-120-686 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-120-687 Supplier Agilent BioTek Supplier No. 02683
Add to Cart
Add to Cart
Flat bottom, polystyrene, sterile, with a lid
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cytation 5, Lionheart FX, Cytation 1, Lionheart LX
Includes Lid
Material Polystyrene
No. of Positions 96
Quantity 5/Pk., 100/Cs.
Sterility Sterile