Learn More
Corning™ Universal Optical Microplate Sealing Tape
GREENER_CHOICE
Catalog No. 05539107 Shop All Corning Products
Change view
Click to view available options
Material:
Polyolefin
Catalog No. Material
05-539-107 Polyolefin
1 options
This item is not returnable. View return policy
Catalog No. 05-539-107 Supplier Corning™ Supplier No. 6575
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pressure-sensitive polyolefin tape ensures tight seal to minimize evaporation
  • Easy application and removal for short- and long-term storage
  • Provides tight seal to minimize evaporation and condensation
  • Aluminum sealing tape (Cat. Nos. 6569 and 6570) is suitable for use between -80°C and 150°C, is not transparent, and is pierceable.
  • Breathable sealing tape (Cat. No. 3345) allows gas exchange across the surface.
  • Universal Optical sealing tape (Cat. No. 6575) is suitable for use between -70°C and 100°C, and is transparent.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polyolefin
Sterility Non-sterile
No. per Case 100/Cs.
No. per Pack 100/Pk.
For Use With (Application) Ideal for microscopic observation of protein crystals, pressure-sensitive tape ensures tight sealing to minimize evaporation and enables oil-free operation when used with thermo cyclers with heated lids
Product Type Optical Microplate Sealing Tape
Temperature Range (Metric) -70°C to +100°C
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov