Learn More
Abcam Coronavirus Protein, Native, Mammalian Cells, ELISA, 1 mg
Description
- Application: ELISA
- Expression System: Mammalian cells
- Formulation: Liquid. Constituents: 0.38% Glycine, 0.1% 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 0.01% Sodium lauryl sulfate
- Storage Conditions: Store at -80°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Protein-based Substrate |
| Target Molecule | Canine Coronavirus |
| Shipping Condition | Dry Ice |
| Label or Dye | Unconjugated |
| Form | Liquid |
| Detection Method | Colorimetric |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. |
| Buffer | 0.38% glycine, 0.1% 3-[(3-cholamidopropyl)dimethylammonio]-1-propanesulfonate, 0.01% sodium lauryl sulfate |
| Formulation | Constituents: 0.38% Glycine, 0.1% 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 0.01% Sodium lauryl sulfate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.