Learn More
Invitrogen™ Coronin 1A Polyclonal Antibody

Description
PA1-10022 was made against the C-terminal peptide of human coronin 1a chemically coupled to keyhole limpet hemocyanin carrier protein. PA1-10022 is also known to work on many mammalian species both on western blots and on cells grown in tissue culture.
Specifications
Specifications
| Antigen | Coronin 1A |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 5mM sodium azide |
| Gene | CORO1A |
| Gene Accession No. | O89053, P31146, Q91ZN1, Q92176 |
| Gene Alias | actin-binding; bovine p57; Clabp; Clabp coronin; CLIPINA; Clipin-A; CORO1; CORO1A; CORO1A protein; coronin 1; coronin 1A; coronin, actin binding protein 1A; coronin, actin binding protein, 1A; coronin-1; coronin-1A; coronin-like actin binding protein; coronin-like protein A; Coronin-like protein p57; FLJ41407; HCORO1; IMD8; Lmb3; MGC117380; p57; TACO; tryptophan aspartate-containing coat protein |
| Gene Symbols | CORO1A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.