Learn More
Cayman Chemical CorosolIc AcId 10mg
SDP
Supplier:  Cayman Chemical 2788710 MG
Encompass_Preferred
A derivative and metabolite of coumarin with diverse biological activities inhibits growth of human ACHN renal A549 lung HCT15 colon and LNCaP prostate carcinoma as well as Dakiki myeloma and HL-60 lymphoma cells at 250-300 Ug/ml increases ROS production and halts the cell cycle in KB human oral carcinoma cells has antidepressant-like activity in a rat model of depression induced by chronic unpredictable mild stress at 15 and 30 mg/kg used as a ratiometric pH indicator with ex/em 330 and 370/460 nm at low and high pH respectively
Catalog No. 50-293-9268
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.