Learn More
New Pig Corrosive Liquids Drum Gauge
SDP
Catalog No. 17987829 Shop All New Pig Corporation Products
Catalog No. 17-987-829 Supplier New Pig Supplier No. DRM184
Add to Cart
Add to Cart
Gauge can be used with flammables and its chemical- and solvent-resistant stainless steel design is built to last
  • Durable stainless steel construction: Made with a chemical- and solvent-resistant stainless steel rod, this gauge is built to withstand harsh, corrosive liquids and provide a long service life.
  • Clear and accurate level readings: The gauges indicator tube shows high and low barrel levels in gallons, liters, and volume percentages, preventing overfilling and helping to manage inventory.
  • Versatile and reliable: Designed for use with a wide range of corrosive and flammable liquids, it provides a simple and effective way to monitor drum contents without messy dipsticks.
  • Minimizes maintenance and downtime: The simple mechanical design is built for durability and minimal upkeep, ensuring reliable performance without frequent repairs.
  • Easy to install: It easily screws into a standard bung opening on a vertical drum, allowing for quick and hassle-free setup.
  • Reduces overflow risks: By providing a clear and visible indication of the liquid level, the gauge helps prevent dangerous and costly spills.
Specifications
Color Silver, Yellow