Learn More
Waters Corp CORTECS C18 Column, 90Å, 2.7 µm, 4.6 mm X 50 mm, 1/pk
Supplier:  Waters Corp 186007375
Encompass_Preferred
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
Catalog No. NC1457396
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.