Learn More
Invitrogen™ Cortical thymocytes Monoclonal Antibody (ICO-44)
Description
MA1-7662 detects Cortical thymocytes from human samples. MA1-7662 has been successfully used in flow cytometry procedures. The MA1-7662 immunogen is Cortical thymocytes.
Specifications
Specifications
| Antigen | Cortical thymocytes |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | ICO-44 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin, 0.5% saccharose and 0.01% sodium azide; pH 7.2 |
| Host Species | Mouse |
| Immunogen | Cortical thymocytes. |
| Purification Method | Protein A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.