Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Corticosterone Polyclonal Antibody

Catalog No. PIPA124900 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA124900 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA124900 Supplier Invitrogen™ Supplier No. PA124900

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Stock solution: Add 5 mL of 0.05 M Tris-HCl buffer, pH 8.0, containing 0.1M NaCl, 0.1% bovine serum albumin and 0.1% sodium azide. Prior to reconstitution store at 4°C. Following reconstitution keep as concentrated solution, aliquot and store at -20°C or below. Avoid freeze-thaw cycles.

CORTICOSTERONE is an adrenocortical steroid that has modest but significant activities as a mineralocorticoid and a glucocorticoid. [National Center for Biotechnology Information. PubChem Compound Database; CID=5753].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Corticosterone
Applications Radioimmune Assays (RIA)
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation TBS with 0.1% BSA; pH 8
Gene Alias 17-Deoxycortisol; C21H30O4
Host Species Rabbit
Immunogen Corticosterone-21-thyroglobulin.
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Lyophilized
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.