Learn More
Cayman Chemical CortIcosTRon ELISA ntIsRu
SDP
Supplier:  Cayman Chemical 401322500 DTN
Encompass_Preferred
A chromone with diverse biological activities inhibits the protein-protein interaction between histone H3 and 14-3-3ε and phosphorylation of histone H3 as well as induces cell cycle arrest at the G2/M phase in HT-29 cells at 10 UM inhibits compound 48/80-induced histamine release in LAD 2 mast cells from 10-100 UM inhibits perivascular adventitia leukocyte infiltration and improves neurological deficits in a rat model of subarachnoid hemorrhage-induced vasospasm at 400 Ug/kg has been used as a quality control marker of S. diviaricata roots
Catalog No. 50-294-2228
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.