Learn More
CPC Scientific H-Ser-Glu-Glu-Pro-Pro-Ile-Ser-Leu-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Met-Ala-Arg-Ala-Glu-Gln-Leu-Ala-Gln-Gln-Ala-His-Ser-Asn-Arg-Lys-Leu-Met-Glu-Ile-Ile-NH2 1MG
SDP
Supplier:  CPC Scientific CRFS001B
Encompass
Hypothalamic hormone that stimulates the synthesis and release of ACTH from the anterior pituitary.ONE-LETTER SEQUENCE: SEEPPISLDLTFHLLREVLEMARAEQLAQQAHSNRKLMEII-NH2MOLECULAR FORMULA: C208H344N60O63S2MOLECULAR WEIGHT:4757.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [86784-80-7], RESEARCH AREA: HormonalREFERENCES: S. Shibahara et al., The EMBO J., 2, 775 (1983)
Catalog No. 50-210-9160