Learn More
Carbosynth LLC. Cortisol-BSA conjugate
SDP
Supplier:  Carbosynth LLC. 801060
Encompass_Preferred
Cortisol is a steroid hormone, more specifically a glucocorticoid, produced by the zona fasciculata of the adrenal cortex. It is released in response to stress and a low level of blood glucocorticoids. Its primary functions are to increase blood sugar through gluconeogenesis; suppress the immune system; and aid in fat, protein and carbohydrate metabolism.
Catalog No. 50-125-3470
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.