Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cortisone 21-acetate, 98+%, Thermo Scientific Chemicals

Catalog No. AC448960050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448960050 5 g Glass bottle
AC448960250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC448960050 Supplier Thermo Scientific Chemicals Supplier No. 448960050
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-04-4
Molecular Formula C23H30O6
Molecular Weight (g/mol) 402.49
MDL Number MFCD00003609
InChI Key ITRJWOMZKQRYTA-RFZYENFJSA-N
Synonym cortisone acetate, cortisone 21-acetate, cortone acetate, incortin, biocort acetate, artriona, cortadren, scheroson, cortisyl, adreson
PubChem CID 5745
ChEBI CHEBI:3897
IUPAC Name [2-[(8S,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3,11-dioxo-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C

Specifications

Melting Point 237°C to 240°C
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,2525
Formula Weight 402.48
Percent Purity 98+%
Chemical Name or Material Cortisone 21-acetate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 200-006-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.