Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cortisone Acetate 99.0+%, TCI America™
SDP

Catalog No. c03891g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03891G 1 g
C038925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C03891G Supplier TCI America Supplier No. C03891G

Chemical Identifiers

CAS 50-04-4
Molecular Formula C23H30O6
Molecular Weight (g/mol) 402.49
MDL Number MFCD00003609
InChI Key ITRJWOMZKQRYTA-RFZYENFJSA-N
Synonym cortisone acetate, cortisone 21-acetate, cortone acetate, incortin, biocort acetate, artriona, cortadren, scheroson, cortisyl, adreson
PubChem CID 5745
ChEBI CHEBI:3897
IUPAC Name 2-[(1R,3aS,3bS,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-7,10-dioxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C

Specifications

Melting Point 238°C
Color White
Quantity 1 g
Formula Weight 402.49
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cortisone Acetate
RTECSNumber GM9140000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.