Learn More
CPC Scientific H-Pro-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Ser-Ser-Cys-Lys-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CORT002B
Encompass
Neuropeptide similar to somatostatin-14 that is mainly expressed in the cortex and shares many functional properties with somatostatin-14 but differs in its sleep modulatory properties.SEQUENCE: H-Pro-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Ser-Ser-Cys-Lys-OH (trifluoroacetate salt)(Cys2 and 13 bridge)ONE-LETTER SEQUENCE: PCKNFFWKTFSSCK (Cys2 and 13 bridge)MOLECULAR FORMULA: C81H113N19O19S2MOLECULAR WEIGHT: 1721.1STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [186901-48-4]SYNONYMS: RESEARCH AREA: NeuropeptidesREFERENCES:L. de Leccea et al., Nature, 381, 242 (1996)S.Fukusumi et al., Biochem. Biophys. Res. Commun., 232, 157 (1997)SKU(s): CORT-002A, CORT-002B
Catalog No. 50-210-9140
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.