Learn More
CPC Scientific Glp-Glu-Arg-Pro-Pro-Leu-Gln-Gln-Pro-Pro-His-Arg-Asp-Lys-Lys-Pro-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Ser-Ser-Cys-Lys-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CORT001B
Encompass
ONE-LETTER SEQUENCE: Glp-ERPPLQQPPHRDKKPCKNFFWKTFSSCK (Cys17 and 28 bridge)MOLECULAR FORMULA: C161H242N46O41S2MOLECULAR WEIGHT:3540.09STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1815618-17-7], SYNONYMS: Cortistatin-29 (rat)RESEARCH AREA: NeuropeptidesREFERENCES: L. de Leccea et al., Nature, 381, 242 (1996)
Catalog No. 50-210-9138
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.