Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products GL601 MicroFoam Nitrile Coated Nylon Gloves

Catalog No. 19555063 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-555-063 Medium
19-555-064 Small
19-555-062 Large
19-555-066 X-Large
19-555-065 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-555-063 Supplier Pyramex Safety Products Supplier No. GL601M
Add to Cart
Add to Cart
This item is not returnable. View return policy

MicroFoam Nitrile coating on the palm and fingers absorbs oil to enhance grip capabilities, tactile sensitivity, and dexterity.

  • Nitrile MicroFoam coating on the palm and fingers enhances grip
  • Additional foam coating acts like a sponge to increase gripping power
  • 15-gauge Nylon Lycra™ shell is tear and abrasion resistant
  • Gloves offer tactile sensitivity and dexterity

Specifications

ANSI Abrasion Rating ANSI 6
Coating Material Nitrile MicroFoam
Color Gray
Hand Specific
Material Nylon, Spandex
Product Type Dip-coated Gloves
Size Medium
For Use With (Application) General Assembly, Automotive Components, Horticulture, and Warehousing
Glove Cuff Style Knit
Series GL601
Yarn Gauge 15
Show More Show Less

WARNING: Cancer and Reproductive Harm - www.P65warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.