Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products GL602C3 Series Micro-Foam Nitrile-Coated HPPE Cut-Resistant Gloves, Gray

Catalog No. 19555067 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-555-067 Large Brown
19-555-069 Small Red
19-555-068 Medium Yellow
19-555-071 X-Large Black
19-555-070 2X-Large Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-555-067 Supplier Pyramex Safety Products Supplier No. GL602C3L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Micro-Foam nitrile coating acts like a sponge to help increase gripping power, while the HPPE shell provides ANSI Level A2 Cut Protection.

  • 13-gauge HPPE shell is resistant to abrasions, cuts, and tears
  • Micro-Foam coating enhances grip capabilities

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Nitrile Foam
Color Gray
Cut Resistance Rating ANSI A2
Hand Specific
Material HPPE
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388:4X42B
For Use With (Application) General Duty, parts assembly, construction, municipal work
Glove Cuff Color Brown
Glove Cuff Style Continuous Knit
Model GL602C3L
Puncture Rating ASTM F2878 LEVEL 3
Series GL602C3
Yarn Gauge 13
Show More Show Less

WARNING: Cancer and Reproductive Harm - www.P65warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.