Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products GL603C5 Series Micro-Foam Nitrile-Coated HPPE Cut-Resistant Gloves, Black

Catalog No. 19555072 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-555-072 Large Brown
19-555-074 Small Red
19-555-073 Medium Yellow
19-555-076 X-Large Black
19-555-075 2X-Large Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-555-072 Supplier Pyramex Safety Products Supplier No. GL603C5L
Add to Cart
Add to Cart
This item is not returnable. View return policy

MicroFoam nitrile coating acts like a sponge to help increase gripping power, while the HPPE shell provides ANSI Level A4 Cut Protection

  • 13-gauge HPPE shell is resistant to abrasions, cuts, and tears
  • MicroFoam coating enhances grip capabilities
  • Dark color hides dirt and stains for extended useful life

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Nitrile Foam
Color Black
Cut Resistance Rating ANSI A4
Hand Specific
Material HPPE
Product Type Cut-resistant Gloves
Size Large
EN388 Standard EN 388:4X43DX
For Use With (Application) Assembly line, furniture manufacturing, HVAC, aerospace and construction
Glove Cuff Color Brown
Glove Cuff Style Continuous Knit
Model GL603C5L
Puncture Rating ASTM F2878 LEVEL 4
Series GL603C5
Yarn Gauge 13
Show More Show Less

WARNING: Cancer and Reproductive Harm - www.P65warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.