Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products GL604C5 Series Sandy Nitrile-Coated HPPE Gloves

Catalog No. 19555079 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-555-079 Small Red
19-555-078 Medium Yellow
19-555-077 Large Brown
19-555-081 X-Large Black
19-555-080 2X-Large Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-555-079 Supplier Pyramex Safety Products Supplier No. GL604C5S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sandy nitrile coating provides enhanced grip capabilities in dry, wet and oily conditions.

  • Sandy nitrile coated gloves
  • 13-gauge HPPE shell is resistant to tears and abrasion
  • Enhances grip capabilities in dry, wet, and oily conditions

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) EN 420
Coating Material Nitrile
Color Gray, Black
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free No
Material HPPE
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:4X43D
For Use With (Application) Assembly lines, manufacturing, HVAC, construction
Glove Cuff Color Red
Glove Cuff Style Elasticated
Grip Design Sandy palm
Model GL604C5S
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Sandy Nitrile GL604C5 Series
Yarn Gauge 13
Show More Show Less

WARNING: Cancer and Reproductive Harm - www.P65warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.