Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Costar™ 384-Well Polypropylene Storage Block

Catalog No. p-156353 Shop All Corning Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
07-200-749 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-200-749 Supplier Corning™ Supplier No. 3964
Only null left
Add to Cart
Add to Cart

Rigid sidewalls for optimal performance in automated handling

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI
Format Standard
Lid Without Lid
Array 16 x 24
Color Clear
Material Polypropylene
No. of Wells 384
Sterility Sterile
Surface Treatment Non-treated
Well Shape Round
For Use With (Application) For high throughput applications requiring added volume
No. per Case 25/Cs.
No. per Pack 5/Pk.
Quantity 25/Cs.
Well Color Clear
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.