Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 96-Well Assay Blocks
GSA_VA

Catalog No. p-156362 Shop All Corning Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Volume (Metric) Well:
500 μL
1000 μL
2000 μL
Well Shape:
Conical Bottom (V-well)
Round
Round, Conical Bottom (V-well)
No. of Wells:
96
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Volume (Metric) Well Well Shape No. of Wells
07-200-701 Non-sterile 2000 μL Conical Bottom (V-well) 96
07-200-724 Non-sterile 500 μL Round, Conical Bottom (V-well) 96
09-761-116A Non-sterile 1000 μL Round 96
07-200-700 Sterile 2000 μL Conical Bottom (V-well) 96
09-761-116 Sterile 1000 μL Round 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07-200-701 Supplier Corning™ Supplier No. 3961
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured from clarified polypropylene for optical clarity. Corning™ 96-Well Assay Blocks are ideal for compound storage, assay, and cell-growth applications.

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile

Certifications

Certified DNase- and RNase-free

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 8 x 12
Packaging 20 inner packs of 5
Binding Type Not Treated
Color Clear
Product Type 96-well Assay Blocks
Material Polypropylene
Volume (Metric) Well 2000 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Conical Bottom (V-well)
For Use With (Application) For compound storage, assay and cell-growth application
No. per Case 100/Cs.
No. per Pack 5/Pk.
Quantity 100/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.