Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cotton Rat CCL3/MIP-1 alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1061
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1061 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1061 Supplier R&D Systems Supplier No. AF1061
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CCL3/MIP-1 alpha Polyclonal specifically detects CCL3/MIP-1 alpha in Cotton Rat samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CCL3/MIP-1 alpha
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.1-0.4 ug/mL
Gene Accession No. AAL26704
Gene Alias C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta
Gene Symbols CCL3
Host Species Goat
Immunogen E. coli-derived recombinant cotton rat CCL3/MIP-1 alpha Ala24-Ala92 Accession # AAL26704
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6348
Test Specificity Detects cotton rat CCL3/MIP-1 alpha in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse MIP-1 alpha is observed, 15% cross-reactivity with recombinant human (rh) MIP-1 alpha is observed, less than 10% cross-reactivity with rhMIP-1 beta , rh6Ckine, and rhHCC-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Cotton Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.