Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumalic Acid 97.0+%, TCI America™
SDP

Catalog No. C039125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C039125G 25 g
C03915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C039125G Supplier TCI America Supplier No. C039125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 500-05-0
Molecular Formula C6H4O4
Molecular Weight (g/mol) 140.09
MDL Number MFCD00006644
InChI Key ORGPJDKNYMVLFL-UHFFFAOYSA-N
Synonym coumalic acid, 2-oxo-2h-pyran-5-carboxylic acid, cumalic acid, 2h-pyran-5-carboxylic acid, 2-oxo, 2-oxopyran-5-carboxylic acid, unii-ob1jpy343g, 2-pyrone-5-carboxylic acid, alpha-pyrone-5-carboxylic acid, .alpha.-pyrone-5-carboxylic acid, ob1jpy343g
PubChem CID 68141
IUPAC Name 2-oxo-2H-pyran-5-carboxylic acid
SMILES OC(=O)C1=COC(=O)C=C1

Specifications

Melting Point 204°C
Boiling Point 218°C
Quantity 25 g
Formula Weight 140.09
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Coumalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.