Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 153 98.0+%, TCI America™
SDP

Catalog No. C28581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28581G 1 g
C2858200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28581G Supplier TCI America Supplier No. C28581G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53518-18-6
Molecular Formula C16H14F3NO2
Molecular Weight (g/mol) 309.288
MDL Number MFCD00041843
InChI Key VSSSHNJONFTXHS-UHFFFAOYSA-N
Synonym coumarin 153, pilot 495, c 6f, coumarin 540a, 1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizin-11-one, 2,3,6,7-tetrahydro-9-trifluoromethyl, 2,3,6,7-tetrahydro-9-trifluoromethyl-1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizin-11-one, 9-trifluoromethyl-2,3,6,7-tetrahydro-1h,5h,11h-pyrano 2,3-f pyrido 3,2,1-ij quinolin-11-one, 8-trifluoromethyl-2,3,5,6-tetrahydro-1h,4h-11-oxa-3a-aza-benzo de anthracen-10-one
PubChem CID 72652
ChEBI CHEBI:51773
SMILES C1CC2=C3C(=C4C(=C2)C(=CC(=O)O4)C(F)(F)F)CCCN3C1

Specifications

Melting Point 166°C
Color Yellow
Quantity 1 g
Formula Weight 309.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 153
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.