Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin-3-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C312925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C312925G 25 g
C31295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C312925G Supplier TCI America Supplier No. C312925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 531-81-7
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00006852
InChI Key ACMLKANOGIVEPB-UHFFFAOYSA-N
Synonym coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 van
PubChem CID 10752
IUPAC Name 2-oxochromene-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O

Specifications

Melting Point 192°C
Quantity 25 g
UN Number 2811
Formula Weight 190.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin-3-carboxylic Acid
RTECSNumber DJ2490000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.