Learn More
Poltex Tilted Countertop Glove Box Holder
SDP
Catalog No. 22000516
Change view
Click to view available options
Capacity:
3 Boxes
4 Boxes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
22-000-516 3 Boxes
22-000-517 4 Boxes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-516 Supplier Poltex Supplier No. TILTGLV3
Only null left
Add to Cart
Add to Cart
Designed with a backward tilt for easy dispensing and stability on any flat surface. Holder is constructed from durable PETG, with rubber bumpons on the base to prevent scratching or slipping.
  • Holder is designed with a backwards tilt for easy dispensing and stability on any flat surface
  • Constructed from durable PETG which features higher chemical resistance than acrylic
  • Rubber bumpons on the base prevent scratching and slipping
  • Top loading design is compatible with virtually any size glove box
  • Gloves are sold separately
Specifications
Product Type Glove Box Holder
Color Clear
Dimensions (D x W x H) Interior 3.5 x 17.5 x 9 in.
Holds Disposable Glove Boxes
Material PETG
No. of Compartments 1
Product Style Top Loading
Capacity 3 Boxes