Learn More
MilliporeSigma™ Supelco™ Coupler for Legacy Guard column Holder, PEEK
Catalog No. 111004843 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
PEEK
Catalog No. Material
11-100-4843 PEEK
1 options
Catalog No. 11-100-4843 Supplier MilliporeSigma™ Supelco™ Supplier No. 54986
Add to Cart
Add to Cart
Finger-tight design couples HPLC columns that use 1/16 in. 10-32 Valco-compatible fittings with seating distance of 0.074 in.. Also used as a replacement for PEEK coupler supplied in stand-alone guard cartridge holder, Cat. No. 54987.
TRUSTED_SUSTAINABILITY
Specifications
Description I.D. O.D. Overall L 0.010 in. 1/16 in. 1 in.
Material PEEK