Learn More
Chemglass Life Sciences 30mm Stirrer Shaft Coupling, For Use With CG-2033-X-11 Motor, Gold Motor Adapter
SDP
Supplier:  Chemglass Life Sciences CG204697
Encompass_Preferred
Coupling incorporates a universal joint to compensate for misalignment of drive motor and glass stirrer shaft. Nylon bushing compresses around shaft using an allen screw. Steel stud attaches directly to the chuck of a stirrer motor. Supplied complete with an allen wrench. PLEASE NOTE: OTHER THAN OUR CG-2097 PROCESS REACTOR STIRRER SHAFTS, CHEMGLASS 19MM OD STIRRER SHAFTS HAVE A REDUCED TOP DIAMETER OF 12. 7MM. WHEN ORDERING A UNIVERSAL COUPLING FOR A CHEMGLASS 19MM OD STIRRER SHAFT, USE PART NUMBER CG-2046-03.
Catalog No. NC2398022
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.