Learn More
Creative Biomart Recombinant CoV-2 S1 (16-685) Protein, His/Avi-tagged, Biotin-Labled
Supplier:  Creative Biomart COV2373
Encompass_Preferred
The HEK293 derived Biotinylated recombinant protein contains the Coronavirus 2019 CoV-2 Spike Glycoprotein S1, Wuhan-Hu-1 strain, amino acids 16-685 fused to His tag & Avi-tag at C-terminal.
Catalog No. 50-208-4751
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.