Learn More
Creative Biomart Recombinant CoV-2-S1 Protein, Fc-tagged
Supplier:  Creative Biomart COV2388
Encompass_Preferred
The HEK293 derived recombinant protein contains the Coronavirus 2019 Spike Glycoprotein S1 Receptor Binding Domain [RBD], Wuhan-Hu-1 strain, amino acids 319-541 fused to Fc tag at C-terminal.
Catalog No. 50-208-4766
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.