Learn More
Creative Biomart Recombinant CoV-2 S2 Protein, Fc-tagged
Supplier:  Creative Biomart COV2377
Encompass_Preferred
The HEK293 derived recombinant protein contains the Novel Coronavirus 2019-nCoV Spike Glycoprotein S2, Wuhan-Hu-1 strain, amino acids 685-1211 fused to Fc tag at C-terminal.
Catalog No. 50-208-4755
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.