Learn More
Med Vet International Dressing Retention Tape With Liner 3M Medipore Dress-It White 7-7/8 X 11 inch Soft Cloth Nonsterile, 2958, 25/Box
Supplier: Med Vet International 683385BX
The 3M Medipore Dress-It pre-cut dressing cover secures dressings where skin may stretch It comes pre-cut to save time and effort The material is gentle and breathable to enhance comfort and maintain the skin It's stretchy to help conform easily around body contours The 7-7/8-inch x 11-inch cloth tape provides no self-adhesion or curling, making it easy to handle The white tape is latex-free to prevent allergic reactions There are 25 non-sterile pads in a box
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.