Learn More
Uniform Technology Coverall Ultimax Grid, White
Catalog No. 17779021 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Large (Long)
3X-Large (Long)
4X-Large (Long)
5X-Large (Long)
6X-Large (Long)
7X-Large
Large (Long)
Medium (Long)
X-Large (Long)
X-Small
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-779-021 5X-Large (Long)
17-779-003 Medium (Long)
17-779-006 Large (Long)
17-779-009 X-Large (Long)
17-779-012 2X-Large (Long)
17-779-015 3X-Large (Long)
17-779-018 4X-Large (Long)
17-010-521 X-Small
17-010-515 7X-Large
17-377-680 6X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 17-779-021 Supplier Uniform Technology Supplier No. CC1245TD09WH5XLL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cleanroom coveralls feature a military collar, raglan sleeves, knit cuffs, and a coil zipper with top snaps.
  • Ideal for General CE Spaces/Applications (Non-ESD sensitive Medical Device, Electronics and Aerospace, Food Processing/Handling or Packaging, Cultivation Industry, University Labs, Imaging/Film coating Manufactures, General Medical/Dental Facilities)
  • Coverall
  • Ultimax Grid
  • Military Collar
  • Raglan Sleeves
  • Knit Cuffs
  • Coil Zipper with Top Snaps
Specifications
Product Type Cleanroom Coveralls
Size 5X-Large (Long)
Gender Unisex
Material Polyester
Color White
Disposable Reusable
Ankle Style Snap
Cuff Style Knit
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Anti-static
Collar Type Military
Latex Free Yes
No. of Pockets 1
Size (Metric) Chest 177.80 cm
Sleeve Style Raglan
Length (Metric) 86.36 cm
Quantity 1/Ea.
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.