Learn More
Uniform Technology Coverall Ultimax Grid, White, Short
Catalog No. 17779001 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-779-001 Medium
17-779-004 Large
17-779-007 X-Large
17-779-010 2X-Large
17-779-013 3X-Large
17-779-016 4X-Large
17-779-019 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-779-001 Supplier Uniform Technology Supplier No. CC1245TD09WHMS
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Short style cleanroom coveralls feature a military collar, raglan sleeves, knit cuffs, and a coil zipper with top snaps.
  • Ideal for General CE Spaces/Applications (Non-ESD sensitive Medical Device, Electronics & Aerospace, Food Processing/Handling or Packaging, Cultivation Industry, University Labs, Imaging/Film coating Manufactures, General Medical/Dental Facilities)
  • Coverall
  • Ultimax Grid
  • Military Collar
  • Raglan Sleeves
  • Knit Cuffs
  • Coil Zipper with Top Snaps
Specifications
Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Polyester
Color White
Disposable Reusable
Ankle Style Snap
Cuff Style Knit
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Anti-static
Collar Type Military
Latex Free Yes
No. of Pockets 1
Size (Metric) Chest 116.84 cm
Sleeve Style Raglan
Length (Metric) 71.12 cm
Quantity 1/Ea.
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.