Learn More
Essendant COVERALLKG A45LGWH
Supplier:  Essendant ANS48973
Encompass_Preferred
Made with lighter weight fabric than the KleenGuard A40 liquid and particle protection suit the A45 suit allows for improved comfort without sacrificing protection Unique Reflex coverall design allows more freedom of movement and less restriction allowing the worker to complete their job without modifying the garment Silicone-free suit is not processed with natural rubber latex making it an excellent choice for employees with allergies Zipper Front Hood and Boots Elastic Wrists and Ankles Apparel Type- Coverall Material(s)- SMS-Based Material Color(s)- White Color Family- White
Catalog No. 50-000-20417
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.