Learn More
Grace Bio Labs Inc COVERWELL PERFUSN CHMBRS,20/PK
Supplier:  Grace Bio Labs Inc 622202
Encompass_Preferred
CoverWell™ Perfusion Chambers.PC1L-2.5, 1-19mm X 32mm X 2.5mm Depth, 26mm X 43mm OD, 1.5mm Diameter Ports – 20 PACK
Catalog No. NC1855191