Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Dermacea™ 100% Cotton Gauze Sponges

An alternative for sterile and nonsterile 100% cotton gauze dressings

Supplier:  Covidien 441218

 View more versions of this product

Catalog No. 22-037-978

Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.

View alternate product


Description

Description

Specifications

Specifications

Cotton
Non-sterile
2 x 2 in.
12-Ply
Gauze Sponge
8000 Cs.
Wound dressing, general cleaning, minor prepping, wound packing and debriding
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit