Learn More
Covidien In-Room™ Sharps Disposal System SharpStar™ Containers
Catalog No. 1483066C
Change view
Click to view available options
Capacity (English):
5 qt.
12 qt.
Color:
Clear
Translucent Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Color
14-830-66C 12 qt. Clear
22-557-177 5 qt. Translucent Red
22-044274 12 qt. Translucent Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-830-66C Supplier Covidien Supplier No. 8536SA
Add to Cart
Add to Cart
This item is not returnable. View return policy
Engineered with controls to prevent overfilling and to limit accidental or intentional access to the container contents
  • Forced horizontal drop maximizes container capacity
  • Anti-kickback feature prevents sharps from exiting the container
  • “FULL” sign signifies that container has reached capacity
  • Locking tabs on top of lid keeps hands away from contents during final closure
  • Molded handles allow safe and secure transport of container
  • Nestable containers save inventory space
Specifications
Capacity (Metric) 11.42 L
Closure Type Counter-Balanced Lid
Capacity (English) 12 qt.
Color Clear
Model Sharpstar
Quantity 10/Cs.
Stackable Yes
Autoclavable Autoclavable