Learn More
Covidien Monoject™ Angel Wing™ Safety Female Transfer Sets
Catalog No. 22029330
Change view
Click to view available options
For Use With (Application):
Blood Transfer
Product Type:
Dome
Tube Holder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Product Type
22-029-330 Blood Transfer Dome
22-225232 Blood Transfer Tube Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-029-330 Supplier Covidien Supplier No. 8881225240
This item is not returnable. View return policy
Safely transfer blood specimens from syringes into blood collection tubes and blood culture bottles
  • Recessed multi-sample needle protected by a plastic shield—easy application of blood collection tube
  • Sets feature needle-shielded holders for transferring blood specimens from syringes
  • Sterile
Cat. No. 22-225-232 Tube Holder is designed for transferring blood from syringes into blood collection tubes; 22-029-330 Dome for transferring blood from syringes into blood culture bottles and blood collection tubes.
Specifications
Product Type Dome
Sterility Sterile
For Use With (Application) Blood Transfer
Includes Holder and Safety Cap