Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Monoject™ Hypodermic Needles with Aluminum Hub

Catalog No. 0556154
Change view
Click to view available options
Needle Gauge:
14 G
16 G
25 G
Length (English) Needle:
1.25 in.
1.5 in.
Needle Point Style:
Bevel
Regular Bevel
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (English) Needle Needle Point Style
05-561-54 25 G 1.25 in. Regular Bevel
05-561-52 16 G 1.5 in. Regular Bevel
22-257-161 14 G 1.5 in. Bevel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-561-54 Supplier Covidien Supplier No. 8881200433

Color-coded, two-piece rigid pack sheath coupled with a precision aluminum hub

  • Ultra-sharp tri-beveled surgical grade stainless steel coated with a special grade of silicone reduces “drag” on the injection site
  • Color-coded needle sheath allows for easy identification of needle gauge
  • Needles are autoclavable, allowing convenient use in hospital-prepared surgical packs
  • Cap-to-sheath seal provides visible and audible tamper evidence and guarantees sterility
  • Rigid polypropylene packaging protects against accidental product contamination

Specifications

Autoclavable Autoclavable
Material Stainless Steel
Needle Gauge 25 G
For Use With (Application) Provides very secure fit, needle can be manipulated with less chance of breakage compared to epoxy deisgn
Includes Precision aluminum hub
Latex Free Yes
Length (English) Needle 1.25 in.
Product Type Hypodermic Needle
Needle Point Style Regular Bevel
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.