Specifications
| For Use With (Application) | Organic Synthesis |
| Type | Cow Type Distillation Receiver |
| Material | Borosilicate Glass |
| For Use With (Equipment) | Distribution adapter |
| Quantity | 1 Ea. |
| Standard Taper Joint | 19/22 |