Learn More
Cayman Chemical COX-2 (mouse) Polyclonal Antibody (aa 584-598), 1 ea
Supplier: Cayman Chemical 1601261
Immunogen: Synthetic peptide from the C-terminal region of mouse COX-2 • Host: Rabbit • Cross Reactivity: (+) Human, macaque monkey, mouse, ovine, and rat COX-2; (−) COX-1 (all species); expected to react with bovine and canine COX-2 • Application(s): IHC (formalin-fixed paraffin-embedded tissue) and WB
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.