Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534813 Supplier Invitrogen™ Supplier No. PA534813
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Molt-4, Mouse brain, Rat brain. Predicted reactivity: Mouse (90%), Rat (88%), Dog (92%), Rabbit (93%), Sheep (93%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PTGS1 (COX1, Cyclooxygenase 1) is the key enzyme in prostaglandin biosynthesis, and acts both as a dioxygenase and as a peroxidase. There are two isozymes of PTGS: a constitutive PTGS1 and an inducible PTGS2, which differ in their regulation of expression and tissue distribution. This gene encodes PTGS1, which regulates angiogenesis in endothelial cells, and is inhibited by nonsteroidal anti-inflammatory drugs such as aspirin. PTGS1 is thought to be involved in cell-cell signaling and maintaining tissue homeostasis. Alternative splicing of this gene generates two transcript variants. The expression of these two transcripts is differentially regulated by relevant cytokines and growth factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PTGS1
Gene Accession No. P22437, P23219, Q63921
Gene Alias COI; COX1; Cox-1; COX3; Cox-3; COXI; cyclooxygenase 3; cyclooxygenase precursor (EC 1.14.99.1); cyclooxygenase-1; Cytochrome c oxidase polypeptide I; Cytochrome c oxidase subunit 1; cytochrome c oxidase subunit I; mitochondrially encoded cytochrome c oxidase I; Mtco1; mt-Co1; P PGG/HS; PCOX1; PES-1; PGG/HS; PGH synthase 1; PGHS1; Pghs-1; PHS 1; PHS1; prostaglandin endoperoxide synthase; prostaglandin G/H synthase 1; prostaglandin G/H synthase and cyclooxygenase; prostaglandin G/H synthase precursor (EC 1.14.99.1); prostaglandin H2 synthase 1; Prostaglandin-endoperoxide synthase 1; prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase and cyclooxygenase); PTGHS; PTGS1; unnamed protein product
Gene Symbols COX1, PTGS1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human COX1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19224, 24693, 4512, 5742
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.