Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX2 Monoclonal Antibody (COX 229), PE-Cyanine7, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 25972582
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-972-582 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-972-582 Supplier Invitrogen™ Supplier No. 25972582
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This COX 229 monoclonal antibody recognizes human COX2 also known as PTGS2. This antibody has been reported to cross-react with the mouse and rat COX2. For optimal performance, we recommend using this antibody with the Foxp3 / Transcription Factor Staining Buffer Set and protocol. Applications Reported: This COX 229 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This COX 229 antibody has been tested by intracellular staining followed by flow cytometric analysis of stimulated normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (Product No. 00-5523-00) and protocol. Please refer to Staining Intracellular Antigens for Flow Cytometry, Protocol B: One step protocol for intracellular (nuclear) proteins located at www.thermofisher.com/flowprotocols. This may be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.

COX2 converts arachidonate to prostaglandin H2 (PGH2), a committed step in prostanoid synthesis, including production of inflammatory prostaglandins. The conversion of arachidonate to prostaglandin H2 is a 2 step reaction: a cyclooxygenase (COX) reaction which converts arachidonate to prostaglandin G2 (PGG2) and a peroxidase reaction in which PGG2 is reduced to prostaglandin H2 (PGH2). It is constitutively expressed in some tissues in physiological conditions, such as the endothelium, kidney and brain, and is up-regulated under pathological conditions, such as in cancer and inflammation (in contrast to the iso-enzyme PTGS1, which is expressed ubiquitously). Up-regulation of COX2 is also associated with increased cell adhesion, phenotypic changes, resistance to apoptosis and tumor angiogenesis. In cancer cells, COX2 is a key step in the production of prostaglandin E2 (PGE2), which plays important roles in modulating motility, proliferation and resistance to apoptosis. COX2 is naturally inhibited by calcitriol (the active form of Vitamin D). Glucocorticoids chronically trans-repress PTGS2 gene activity in vivo in part by interfering with transcription initiation and elongation. COX2 is a target of NSAID such as aspirin, which can reduce pain and swelling from inflammation driven by COX2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX2
Applications Flow Cytometry
Classification Monoclonal
Clone COX 229
Concentration 0.2 mg/mL
Conjugate PE-Cyanine7
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene PTGS2
Gene Accession No. P35354
Gene Alias COII; COX2; Cox-2; COXII; cyclooxygenase; cyclooxygenase 2; cyclooxygenase 2b; cyclooxygenase-2; Cytochrome c oxidase polypeptide II; Cytochrome c oxidase subunit 2; cytochrome c oxidase subunit II; glucocorticoid-regulated inflammatory cyclooxygenase; Gripghs; hCox-2; Macrophage activation-associated marker protein P71/73; mitochondrially encoded cytochrome c oxidase II; MTCO2; MT-CO2; PES-2; PGG/HS; PGH synthase 2; Pghs2; PGHS-2; Pghs-b; PHS II; PHS-2; prostaglandin G/H synthase 2; Prostaglandin H2 synthase 2; prostaglandin-endoperoxide synthase 2; prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase and cyclooxygenase); Ptgs2; Tis10; TIS10 protein
Gene Symbols PTGS2
Host Species Mouse
Immunogen Synthetic peptide from human COX2.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5743
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.