Learn More
Invitrogen™ COX2 Recombinant Rabbit Monoclonal Antibody (RM348)

Description
Specificity: reacts human COX-2 (Cyclooxygenase-2) (Prostaglandin G/H synthase 2). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Specifications
Specifications
| Antigen | COX2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | RM348 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide |
| Gene | PTGS2 |
| Gene Accession No. | P35354, Q05769 |
| Gene Alias | COII; COX2; Cox-2; COXII; cyclooxygenase; cyclooxygenase 2; cyclooxygenase 2b; cyclooxygenase-2; Cytochrome c oxidase polypeptide II; Cytochrome c oxidase subunit 2; cytochrome c oxidase subunit II; glucocorticoid-regulated inflammatory cyclooxygenase; Gripghs; hCox-2; Macrophage activation-associated marker protein P71/73; mitochondrially encoded cytochrome c oxidase II; MTCO2; MT-CO2; PES-2; PGG/HS; PGH synthase 2; Pghs2; PGHS-2; Pghs-b; PHS II; PHS-2; prostaglandin G/H synthase 2; Prostaglandin H2 synthase 2; prostaglandin-endoperoxide synthase 2; prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase and cyclooxygenase); Ptgs2; Tis10; TIS10 protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.