Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ COX4 Antibody Blocking Peptide
SDP

Catalog No. NB398641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398641 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398641 Supplier Novus Biologicals™ Supplier No. NB11039115PEP
Add to Cart
Add to Cart

Specifications

Gene ID (Entrez) 1327
Species Human
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB110-39115
Gene Alias COX IV-1, CO x 4-1, CO x 4 COXIV, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1 cytochrome c oxidase subunit IV, MGC72016
Gene Symbol COX4I1
Quantity 0.1 mg
Research Category Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism
Storage Buffer Peptide dissolved in dH2O. Contains no BSA.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.